7-Hydroxyflavone 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one

OR1155-25G OR1155-5G OR1155-1G
76565-298EA 347.9 USD
76565-298 76565-296 76565-294
7-Hydroxyflavone 95%
7-Hydroxyflavone
Formula: C₁₅H₁₀O₃
MW: 238.24 g/mol
Boiling Pt: 395
Melting Pt: 243…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00006835
CAS Number: 6665-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR