2-Acetylamino-5-bromo-6-methylpyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Acetamido-5-bromo-6-methylpyridine, 6-Acetamido-3-bromo-2-methylpyridine

OR11546-5G OR11546-25G OR11546-1G
76565-290EA 46.86 USD
76565-290 76565-292 76565-288
2-Acetylamino-5-bromo-6-methylpyridine 98%
2-Acetylamino-5-bromo-6-methylpyridine
Formula: C₈H₉BrN₂O
MW: 229.0700 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD03909755
CAS Number: 142404-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR