2-Bromo-1-(o-tolyl)ethanone

Supplier: APOLLO SCIENTIFIC
Warning

OR11269-5G OR11269-1G
76565-044EA 273.98 USD
76565-044 76565-042 76565-040
2-Bromo-1-(o-tolyl)ethanone
2-Bromo-1-(o-tolyl)ethanone
Formula: C₉H₉BrO
Boiling Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD04038955
CAS Number: 51012-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR