Pyrogallol 98+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene

OR11265-1KG OR11265-500G OR11265-100G
76565-038EA 144.66 USD
76565-038 76565-036 76565-034
Pyrogallol 98+%
Pyrogallol
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 309
Melting Pt: 131…135 °C
Density: 1.45 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002192
CAS Number: 87-66-1
EINECS: 201-762-9
Merck Index: 13,08090

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR