6-Benzylaminopurine

Supplier: APOLLO SCIENTIFIC
Warning

OR11257-5G OR11257-25G OR11257-100G
76565-026EA 20.77 USD
76565-026 76565-028 76565-030
6-Benzylaminopurine
6-Benzylaminopurine
Formula: C₁₂H₁₁N₅
Melting Pt: 229…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00005572
CAS Number: 1214-39-7
EINECS: 214-927-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR