5-(Hydroxymethyl)-4-(4-methoxyphenyl)-2-(propan-2-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one

Supplier: APOLLO SCIENTIFIC

OR111518-250MG OR111518-1G
76564-882EA 166.04 USD
76564-882 76564-884
5-(Hydroxymethyl)-4-(4-methoxyphenyl)-2-(propan-2-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
5-(Hydroxymethyl)-4-(4-methoxyphenyl)-2-(propan-2-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Formula: C₁₃H₁₇N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD31629932
CAS Number: 2244086-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR