6-Chloroquinoxaline

Supplier: APOLLO SCIENTIFIC
Danger

OR11146-25G OR11146-5G OR11146-1G
76564-850EA 88.62 USD
76564-850 76564-848 76564-846
6-Chloroquinoxaline
6-Chloroquinoxaline
Formula: C₈H₅ClN₂
MW: 164.5917 g/mol
Boiling Pt: 117…119 °C
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00047594
CAS Number: 5448-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR