Ethyl 4-bromo-5-methyl-1H-pyrazole-3-carboxylate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR111144-5G
OR111144-1G
76564-702EA
556.2
USD
76564-702
76564-700
76564-698
Ethyl 4-bromo-5-methyl-1H-pyrazole-3-carboxylate
Ethyl 4-bromo-5-methyl-1H-pyrazole-3-carboxylate
Formula:
C₇H₉BrN₂O₂ Melting Pt: 131…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD11054303 CAS Number: 6076-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...