3-Bromo-o-xylene 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1-Bromo-2,3-dimethylbenzene
OR11088-500G
OR11088-100G
OR11088-25G
76564-474EA
477.34
USD
76564-474
76564-472
76564-470
3-Bromo-o-xylene 99%
3-Bromo-o-xylene
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 214 Density: 1.365 g/cm³ (25 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000069 CAS Number: 576-23-8 EINECS: 209-398-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...