5-Aminobenzimidazole

Supplier: APOLLO SCIENTIFIC
Warning

OR110728-5G OR110728-1G
76564-322EA 45.84 USD
76564-322 76564-320
5-Aminobenzimidazole
5-Aminobenzimidazole
Formula: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD00465258
CAS Number: 934-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR