4-Methylbenzophenone 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR11067-250G OR11067-50G OR11067-1KG
76564-256EA 41.42 USD
76564-256 76564-254 76564-258
4-Methylbenzophenone 99%
4-Methylbenzophenone
Formula: C₁₄H₁₂O
Boiling Pt: 326 °C
Melting Pt: 56.5…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00008553
CAS Number: 134-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR