2-(4-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole

Supplier: APOLLO SCIENTIFIC

OR110416-1G OR110416-5G
76564-106EA 128.36 USD
76564-106 76564-108
2-(4-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
2-(4-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
Formula: C₉H₆BrClN₂O
Melting Pt: 187…188 °C
Storage Temperature: Ambient
MDL Number: MFCD03273472
CAS Number: 568544-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR