N-Methylthiophene-2-sulfonamide

Supplier: APOLLO SCIENTIFIC

OR110294-5G OR110294-1G OR110294-250MG
76562-288EA 686.58 USD
76562-288 76561-088 76562-244
N-Methylthiophene-2-sulfonamide
N-Methylthiophene-2-sulfonamide
Formula: C₅H₇NO₂S₂
MW: 177.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00975951
CAS Number: 53442-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR