3-Chloro-5-methylimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR110129-1G OR110129-250MG
76563-688EA 335.28 USD
76563-688 76563-686
3-Chloro-5-methylimidazo[1,2-a]pyridine
3-Chloro-5-methylimidazo[1,2-a]pyridine
Formula: C₈H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD21336365
CAS Number: 334711-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR