3,5-Dimethylphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid

OR10931-25G OR10931-100G OR10931-5G
76563-520EA 35.66 USD
76563-520 76563-522 76563-518
3,5-Dimethylphenylboronic acid
3,5-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 235…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00185689
CAS Number: 172975-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR