2-Indancarboxylic acid 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3-Dihydro-1H-indene-2-carboxylic acidIndan-2-carboxylic acid

OR10885-5G OR10885-250MG OR10885-1G
76563-474EA 151.41 USD
76563-474 76563-472 76563-470
2-Indancarboxylic acid 95+%
2-Indancarboxylic acid
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00085095
CAS Number: 25177-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR