(Methoxymethyl)triphenylphosphonium bromide 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10663-1G OR10663-5G
76562-022EA 44.82 USD
76562-022 76562-206
(Methoxymethyl)triphenylphosphonium bromide 98%
(Methoxymethyl)triphenylphosphonium bromide
Formula: C₂₀H₂₀BrOP
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00075442
CAS Number: 33670-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR