3-tert-Butyl-1-bromobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR1066-100G OR1066-25G OR1066-5G
76563-240EA 288.28 USD
76563-240 76562-198 76562-240
3-tert-Butyl-1-bromobenzene 99%
3-tert-Butyl-1-bromobenzene
Formula: C₁₀H₁₃Br
Boiling Pt: 95…98 °C/10mmHg
Density: 1.251 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00234661
CAS Number: 3972-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR