Polyethylene glycol 1,000
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
PEG 1,000
, PEG 900-1100
OR1064-50G
76562-996EA
63
USD
76562-996
Polyethylene glycol 1,000
Polyethylene glycol 1,000
Formula:
H(OCH₂CH₂)nOH MW: 1000 g/mol Melting Pt: 61.7…66.6 °C Density: 1.21 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...