2-Amino-6-chloropurine 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR10600-100G OR10600-25G OR10600-500G
76559-942EA 30.56 USD
76559-942 76562-976 76562-400
2-Amino-6-chloropurine 99%
2-Amino-6-chloropurine
Formula: C₅H₄ClN₅
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00075252
CAS Number: 10310-21-1
EINECS: 233-686-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR