2-Amino-5-nitrothiazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine

OR10323-100G OR10323-1KG OR10323-500G
76559-616EA 23.26 USD
76559-616 76562-360 76561-498
2-Amino-5-nitrothiazole 97%
2-Amino-5-nitrothiazole
Formula: C₃H₃N₃O₂S
MW: 145.14 g/mol
Melting Pt: 203 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005326
CAS Number: 121-66-4
EINECS: 204-490-9
Merck Index: 13,00456

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR