4-Amino-2-chloro-6,7-dimethoxyquinazoline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline

OR10316-25G OR10316-5G
76559-784EA 24.01 USD
76559-784 76559-470
4-Amino-2-chloro-6,7-dimethoxyquinazoline
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Formula: C₁₀H₁₀ClN₃O₂
MW: 239.66 g/mol
Melting Pt: 262…268 °C
Storage Temperature: Ambient
MDL Number: MFCD00051734
CAS Number: 23680-84-4
EINECS: 245-821-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR