2-Bromo-5-methylpyridine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR0845-100G
OR0845-25G
76560-972EA
41.76
USD
76560-972
76559-678
2-Bromo-5-methylpyridine
2-Bromo-5-methylpyridine
Formula:
C₆H₆BrN Boiling Pt: 95…96 °C/12.5mmHg Melting Pt: 40…45 °C Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00209553 CAS Number: 3510-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...