4-(Boc-amino)piperidine 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine
OR0807-100G
OR0807-25G
76561-216EA
89.64
USD
76561-216
76562-298
4-(Boc-amino)piperidine 97%
4-(Boc-amino)piperidine
Formula:
C₁₀H₂₀N₂O₂ Melting Pt: 162…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00798171 CAS Number: 73874-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...