Tetrabutylammonium chloride

Supplier: APOLLO SCIENTIFIC
Warning

OR01703-500G OR01703-1KG OR01703-100G
76562-932EA 78.28 USD
76562-932 76561-270 76560-274
Tetrabutylammonium chloride
Tetrabutylammonium chloride
Formula: C₁₆H₃₆ClN
MW: 277.92 g/mol
Melting Pt: 70 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011635
CAS Number: 1112-67-0
EINECS: 214-195-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR