(S)-(+)-2-Aminobutyric acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: L-2-Aminobutyric acidL-alpha-Amino-N-Butyric acid(S)-2-Aminobutanoic acidH-Abu-OH

OR01636-100G OR01636-5G OR01636-25G
76561-394EA 14.35 USD
76561-394 76561-958 76561-684
(S)-(+)-2-Aminobutyric acid
(S)-(+)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064415
CAS Number: 1492-24-6
EINECS: 216-083-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR