2-Ethynylbenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Formylphenylacetylene

OR01514-1G OR01514-5G
76562-676EA 33.62 USD
76562-676 76562-774
2-Ethynylbenzaldehyde 97%
2-Ethynylbenzaldehyde
Formula: C₉H₆O
MW: 130.143 g/mol
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD04039985
CAS Number: 38846-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR