4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR015022-1G OR015022-250MG OR015022-100MG
76562-424EA 271.98 USD
76562-424 76560-352 76560-198
4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane
Formula: C₉H₁₇BO₃
MW: 184.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26521383
CAS Number: 1396215-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR