3-Acetylbenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3'-Acetophenonecarboxylic acid, 3-Acetylbenzoic Acid

OR0060-100G OR0060-25G OR0060-5G
76560-246EA 245.5 USD
76560-246 76560-272 76560-266
3-Acetylbenzoic acid 98%
3-Acetylbenzoic acid
Formula: C₉H₈O₃
Melting Pt: 163…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00045847
CAS Number: 586-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR