3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3,5-Di-tert-butylsalicylaldehyde
OR0037-100G
OR0037-25G
OR0037-500G
76562-184EA
23.38
USD
76562-184
76561-016
76560-440
3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde
Formula:
C₁₅H₂₂O₂ MW: 234.34 g/mol Melting Pt: 59…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191998 CAS Number: 37942-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...