2-Chloro-4-methoxyphenol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Chlor-4-hydroxyanisole

OR0024-25G OR0024-5G
76561-634EA 28.44 USD
76561-634 76559-468
2-Chloro-4-methoxyphenol
2-Chloro-4-methoxyphenol
Formula: C₇H₇ClO₂
MW: 158.58 g/mol
Boiling Pt: 242
Melting Pt: 39…44 °C
Density: 1.153 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00070773
CAS Number: 18113-03-6
EINECS: 242-007-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR