1-Chloro-4-iodobenzene

Supplier: APOLLO SCIENTIFIC
Warning

OR0022-100G OR0022-1KG OR0022-500G
76559-534EA 29.23 USD
76559-534 76560-620 76559-628
1-Chloro-4-iodobenzene
1-Chloro-4-iodobenzene
Formula: C₆H₄ClI
MW: 238.45
Boiling Pt: 226
Melting Pt: 57
Density: 1.886
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001053
CAS Number: 637-87-6
EINECS: 211-305-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR