5-Bromo-2-chlorophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0018-100G OR0018-25G OR0018-5G
76560-808EA 173.18 USD
76560-808 76561-090 76560-288
5-Bromo-2-chlorophenol 98%
5-Bromo-2-chlorophenol
Formula: C₆H₄BrClO
Boiling Pt: 234 °C
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00672940
CAS Number: 183802-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR