Bis(4-chlorophenyl) disulfide 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Bis(4-chlorophenyl) disulphide, Bis(4-chlorophenyl) disulfide

OR0016-10G OR0016-50G
76563-332EA 24.46 USD
76563-332 76559-796
Bis(4-chlorophenyl) disulfide 98%
Bis(4-chlorophenyl) disulphide
Formula: C₁₂H₈Cl₂S₂
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00013642
CAS Number: 1142-19-4
EINECS: 214-531-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR