2-(4-Aminophenyl)ethanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Aminophenethyl alcohol

OR0014-100G OR0014-25G
76562-704EA 67.24 USD
76562-704 76560-066
2-(4-Aminophenyl)ethanol
2-(4-Aminophenyl)ethanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 255 °C
Melting Pt: 108
Storage Temperature: Refrigerator
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR