2'-Aminoacetanilide

Supplier: APOLLO SCIENTIFIC
Warning

OR0008-5G OR0008-100G OR0008-25G
76561-310EA 20.26 USD
76561-310 76560-890 76562-108
2'-Aminoacetanilide
2'-Aminoacetanilide
Formula: C₈H₁₀N₂O
MW: 150.18
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00210388
CAS Number: 34801-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR