Tin (IV) sulfide 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tin disulfide, Tin disulphide

IN3676-10G IN3676-5G
76563-144EA 179.28 USD
76563-144 76562-212
Tin (IV) sulfide 99,9%
Tin (IV) sulphide
Formula: S₂Sn
MW: 182.84
Boiling Pt: 600 °C
Melting Pt: 600
Density: 4.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049546
CAS Number: 1315-01-1
Merck Index: 12,08936

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR