Palladium(II) acetate 99.95+%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
IN3435-1G
IN3435-5G
76560-756EA
69.26
USD
76560-756
76561-654
Palladium(II) acetate 99.95+%
Palladium (II) acetate
Formula:
C₄H₆O₄Pd Melting Pt: 216.3…223.7 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012453 CAS Number: 3375-31-3 EINECS: 222-164-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...