Potassium iodide

Supplier: APOLLO SCIENTIFIC
Warning

IN2918-1KG IN2918-250G
76562-226EA 172.16 USD
76562-226 76561-160
Potassium iodide
Potassium iodide
Formula: Kl
Boiling Pt: 1330 °C
Melting Pt: 681 °C
Density: 3.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR