Potassium bromide 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

IN2891-100G IN2891-25G
76561-946EA 61.12 USD
76561-946 76563-068
Potassium bromide 99,9%
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR