Palladium(II) chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Palladium dichloride

IN2804-250MG IN2804-5G IN2804-1G
76561-644EA 234 USD
76561-644 76561-350 76560-660
Palladium(II) chloride 98%
Palladium (II) chloride

  • Less volume needed than for ethanol precipitation
  • Useful for precipitations with large sample volumes

Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678…680 °C
Density: 4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR