Manganese(II) chloride tetrahydrate 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Manganese chloride tetrahydrate
, Manganous chloride tetrahydrate
, Manganese Chloride, Tetrahydrate
, Manganese(II) chloride tetrahydrate
, Manganese dichloride tetrahydrate
, Manganese chloride (tetrahydrate)
IN2521-500G
IN2521-100G
IN2521-1KG
76562-960EA
84
USD
76562-960
76561-086
76559-592
Manganese(II) chloride tetrahydrate 99%
Manganese (II) chloride tetrahydrate
Formula:
MnCl₂·4H₂O MW: 197.9 g/mol Boiling Pt: 106 °C Melting Pt: 58 °C Density: 2.01 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149792 CAS Number: 13446-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...