Ammonium tetrathiomolybdate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ammonium tetrathiomolybdate(VI)

IN1087-10G IN1087-50G
76562-914EA 289.3 USD
76562-914 76563-090
Ammonium tetrathiomolybdate
Ammonium tetrathiomolybdate
Formula: H₁₀MoN₂S₄
MW: 262.284 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00136013
CAS Number: 15060-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR