N,N-Dimethylformamide-[D₇] (99.5% D) for NMR spectroscopy

Supplier: VWR International
Warning

Synonyms: N,N-Di[2H3]methyl[2H]formamideDMF-D7DMF-d7N,N-Dimethylformamide-[²H₇]Formic acid dimethylamide-D7Heptadeutero-N,N-dimethylformamideFormdimethylamide-D7N,N-Dimethylformamide-d7

300506 301945 302029 301420 300727 300728
BDH0914-0.75MLEA 374.99 USD
BDH0914-0.75ML BDH0914-0.6ML BDH0914-10ML BDH0914-25ML BDH0914-100ML BDH0914-500ML
N,N-Dimethylformamide-[D₇] (99.5% D) for NMR spectroscopy
N,N-Dimethylformamide-[D7]

Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).


  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.

Formula: DCOON(CD₃)₂
MW: 80.04 g/mol
Boiling Pt: 153 °C (1,013 hPa)
Melting Pt: –60 °C (1,013 hPa)
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3,III

Order Now

Specification Test Results

Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.50 %(mol)
w(H2O) 1H-NMR max. 0.030 %(w)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.050 %(w)
GC Content min. 99.0 %(w)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR