Methanol-[D₄] (99.8% D) for NMR spectroscopy

Supplier: VWR International
Danger

BDH0913-25ML BDH0913-10ML BDH0913-100ML
BDH0913-25MLEA 105.4 USD
BDH0913-25ML BDH0913-10ML BDH0913-100ML
Methanol-[D₄] (99.8% D) for NMR spectroscopy
Methanol-[D4]

Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).


  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.

Formula: D₃COD
Boiling Pt: 64.7 … 65.4 °C (1,013 hPa)
Melting Pt: –97.8 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 9.7 °C (1,013 hPa) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3(6.1),II

Order Now

Specification Test Results

Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.80 %(mol)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.030 %(w)
GC Content min. 99.0 %(w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR