Dimethyl sulfoxide-[D₆] (99.8% D) + 0.03% TMS for NMR spectroscopy

Supplier: VWR International

BDH0910-10ML BDH0910-25ML
BDH0910-10MLEA 42.78 USD
BDH0910-10ML BDH0910-25ML
Dimethyl sulfoxide-[D₆] (99.8% D) + 0.03% TMS for NMR spectroscopy
Dimethyl sulphoxide-[D6]

Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).


  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.

Formula: D₃CS(O)CD₃
Boiling Pt: ≥173 … ≤209 °C (1,013 mbar)
Melting Pt: ≥18 … ≤25 °C (1,013 mbar)
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 88 °C (1 atm) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.80 %(mol)
w(H2O) 1H-NMR max. 0.020 %(w)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.030 %(w)
v(TMS) 1H-NMR 0.02-0.04 %V)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR