Deuterium oxide (99.95% D) for NMR spectroscopy
Supplier: VWR International
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).
- Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.
Formula:
D₂O Boiling Pt: 101 °C Melting Pt: 3.81 °C Density: 1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00044636 CAS Number: 7789-20-0 EINECS: 232-148-9 |
Specification Test Results
Appearance/color | Colorless |
Appearance/form | Liquid |
D content 1H-NMR | min. 99.95 %(mol) |
1H-NMR Purity | conform |
TRIT | max.74 Bq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...