Chloroform-[D₁] Isotope_(99.8% D) NMR
Supplier: VWR International
|
Danger
|
Synonyms:
Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
BDH0904-100ML
BDH0904-10ML
BDH0904-25ML
BDH0904-100MLEA
52.88
USD
BDH0904-100ML
BDH0904-10ML
BDH0904-25ML
Chloroform-[D₁] Isotope_(99.8% D) NMR
Chloroform-[D1]
Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).
- Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.
Formula:
CDCl₃ MW: 120.38 g/mol Boiling Pt: 60.8 Melting Pt: -64.1 Density: 1.45 g/cm³ (20 °C) Flash Pt: > 60 °C (1019 hPa) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000827 CAS Number: 865-49-6 EINECS: 212-742-4 UN: 1888 ADR: 6.1 III |
Specification Test Results
Appearance/color | Colorless |
Appearance/form | Liquid |
D content 1H-NMR | min. 99.80 %(mol) |
w(H2O) 1H-NMR | max. 0.010 %(w) |
1H-NMR Purity | conform |
H2O + D2O Content by KF Coulometer | max 0.020 %(w) |
GC Content | min. 99.0 %(w) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...