Acetone-[D₆] (99.8% D) for NMR spectroscopy

Supplier: VWR International
Danger

Synonyms: Propanone-D6DMK-D6HexadeuteroacetoneAcetone-d6Dimethyl ketone-D6Acetone-[²H₆]

300814 300820 300531 300364 300484
BDH0901-0.6MLEA 45.83 USD
BDH0901-0.6ML BDH0901-0.75ML BDH0901-25ML BDH0901-100ML BDH0901-10ML
Acetone-[D₆] (99.8% D) for NMR spectroscopy
Acetone-[D6]

Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).


  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.

Formula: D₃CC(O)CD₃
MW: 64.03 g/mol
Boiling Pt: 56.1 °C (1013 hPa)
Melting Pt: –94.8 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: –17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Order Now

Specification Test Results

Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.80 %(mol)
w(H2O) 1H-NMR max. 0.020 %(w)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.030 %(w)
GC Content min. 99.0 %(w)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR