Ohmeda Infant Warmers
Supplier: GUMBO MEDICAL
O3300IW O4400IW
470346-550EA 3121.89 USD
470346-550 470346-650
Ohmeda Infant Warmers
Incubators Infant Incubators
For delivery rooms or surgery.
  • Refurbished
  • 1 year parts warranty
  • For educational and training use only
A microprocessor-based servo-controlled infant radiant warmer that incorporates a tilting bassinet with folding side panels, an examination lamp, and an overhead infrared radiant heater with digital display and elapsed timer. The 3300 incorporates a fixed position bed, and the 4400 incorporates an elevating bed.
Delivery information: This item has to ship by freight, Additional shipping charges may apply.
Caution: This product is refurbished and is intended for educational and training use only.